Learn More
CD200 Monoclonal Antibody (OX90), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA170101
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-70101 detects CD200 in mouse samples. MA1-70101 has been successfully used in flow cytometry, ELISA, and immunohistochemistry (frozen sections) procedures. The MA1-70101 immunogen is mouse CD200-rat CD4 fusion protein. Store at 4oC short term. For extended storage aliquot and store at -20oC or below. Avoid freeze-thaw cycles.
CD200 is a type-1 membrane glycoprotein that contains two immunoglobulin domains, and belongs to the immunoglobulin superfamily. CD200 is a broadly distributed cell surface glycoprotein that interacts with a receptor on myeloid cells (CD200R) and is implicated in locally preventing macrophage activation. Distant, but recognizable, homologues of CD200 have been identified in many herpesviruses and poxviruses. Studies of the related genes in mouse and rat suggest that CD200 may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms of CD200 have been found. Diseases associated with CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy.Specifications
CD200 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O54901 | |
Cd200 | |
Mouse CD200-rat CD4 fusion protein. | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen) | |
OX90 | |
Unconjugated | |
Cd200 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
Rat | |
Protein G | |
RUO | |
17470 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |