missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD200 Monoclonal Antibody (OX104), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£154.00 - £340.00

Specifications

Antigen CD200
Clone OX104
Concentration 0.6 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30056256
View Documents
Invitrogen™
H038T03Y03-A
100 Tests
£340.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30054821
View Documents
Invitrogen™
H038T02Y03-A
25 Tests
£154.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD200
0.6 μg/Test
Monoclonal
Mouse
Human
P41217
4345
Primary
4°C, store in dark, DO NOT FREEZE!
OX104
Flow Cytometry
NovaFluor Yellow 610
RUO
PBS with 0.09% sodium azide; pH 7.2
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein
IgG1 κ
Affinity chromatography
Cd200
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.