Learn More
CD20 Rabbit Polyclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments with Epredia™ CD20, Rabbit Polyclonal Antibody.
£187.00 - £992.00
Specifications
| Antigen | CD20 |
|---|---|
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12673297
|
Epredia
RB-9013-P0 |
100 μL | Concentrated | ||||||
|
12683297
|
Epredia
RB-9013-P1 |
500 μL | Concentrated | ||||||
|
12663297
|
Epredia
RB-9013-P |
1 mL | Concentrated | ||||||
|
12603307
|
Epredia
RB-9013-R7 |
7 mL | Prediluted | ||||||
Description
CD20 is a 33kDa leukocyte surface antigen consisting of four transmembrane regions and cytoplasmic N- and C-termini. CD20 is expressed primarily on B cells but has also been detected on both normal and neoplastic T cells. Activation of CD20 significantly increases DNA synthesis and is thought to involve basic helix-loop-helix leucine zipper transcription factors.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: Synthetic peptide derived from C-terminus of human CD20 protein
Molecular Weight: 33-37kDa
Positive Control: Tonsil
Cellular Localization: Membrane/cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
| CD20 | |
| Polyclonal | |
| Rabbit | |
| B-Cell | |
| Synthetic peptide derived from C-terminus of human CD20 protein | |
| Primary |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.