missing translation for 'onlineSavingsMsg'
Learn More

CD20 Mouse anti-Human, Clone: B-H20, Novus Biologicals™

Product Code. 18367395 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367395 500 μg 500µL
18308856 200 μg 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18367395 Supplier Novus Biologicals Supplier No. NBP314594500UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD20
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone B-H20
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Functional
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Host Species Mouse
Immunogen Chronic Lymphoid Leukaemia cells (CLL)
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 931
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.