missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD20 Monoclonal Antibody (2H7), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£160.00 - £340.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Concentration | 0.4 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055554
|
Invitrogen™
H076T03Y04-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055036
|
Invitrogen™
H076T02Y04-A |
25 Tests |
£160.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow CytometrySpecifications
| CD20 | |
| 0.4 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P11836 | |
| 931 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 2H7 | |
| Flow Cytometry | |
| NovaFluor Yellow 660 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AA960661; APY; ATOPY; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; High affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; LEU16; LEU-16; Leukocyte surface antigen Leu-16; Ly44; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
| IgG2b κ | |
| Affinity chromatography | |
| Ms4a1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title