missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD20 Antibody Pair

Product Code. 18347807 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347807 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347807 Supplier Novus Biologicals™ Supplier No. H00000931AP22

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.03 ng/mL to 100 ng/mL
Conjugate Unconjugated
Product Type Sandwich ELISA
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 931
Gene Symbol MS4A1
Quantity 1 Set
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Synonym B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Target Human
Buffer 1x PBS, pH 7.4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.