missing translation for 'onlineSavingsMsg'
Learn More

CD20 Antibody (MS4A1/6993R), Novus Biologicals™

Rabbit Monoclonal Antibody

£205.00 - £377.00

Specifications

Antigen CD20
Clone MS4A1/6993R
Concentration 0.2 mg/mL
Dilution Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18663894
View Documents
Novus Biologicals
NBP3-23745-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18623125
View Documents
Novus Biologicals
NBP3-23745-20ug
20 μg
£205.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

CD20
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
10 mM PBS with 0.05% BSA
931
IgG κ
Protein A purified
MS4A1/6993R
Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Recombinant fragment (around aa 97-297) corresponding to C-terminal of human CD20 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.