missing translation for 'onlineSavingsMsg'
Learn More

CD20 Antibody (IGEL/1497R), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP2-49872-0.1mg

 View more versions of this product

Product Code. 18645477

  • £435.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD20
Monoclonal
0.2 mg/mL
Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL
P11836, P11836
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
IGEL/1497R
Unconjugated
10 mM PBS with 0.05% BSA
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Recombinant human full-length CD20 protein (Uniprot: P11836)
0.1 mg
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
931
Store at 4°C.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.