missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD20 Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry, immunofluorescence and flow cytometry applications.

Brand:  Epredia™ MS-340-S0

Code : LD

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12651067

  • £112.00 / 100µL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Mouse

Clone: L26

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with cow, dog, pig and rat.

Epitope: Cytoplasmic domain

Immunogen: Human tonsil B cells

Molecular Weight: 30-33kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
Specifications

Specifications

CD20 Ab-1
Monoclonal
Unconjugated
Human tonsil B cells
IVD
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Paraffin)
L26
Mouse
100 μL
B-Cell
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers