missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Mouse anti-Human, Clone: TP1/31 (CF-Blue), Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13676.100Reactions
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human CD2.
- High specificity
- High reactivity
Specifications
| CD2 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| P06729 | |
| CD2 | |
| Normal human peripheral blood lymphocytes. | |
| 100 Reactions | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| TP1/31 | |
| Mouse monoclonal antibody raised against human CD2. | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| SRBC/T11 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 914 | |
| Store in the dark at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction