missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD2/CD20 Antibody Cocktail, FITC, PE
Antibody Cocktail
£414.00
Specifications
| Antigen | CD2/CD20 |
|---|---|
| Clone | T6.3, DFA-7, DFA-7 |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Classification | Cocktail |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11523533
|
Invitrogen™
MA112058 |
100 Tests |
£414.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11533533
|
Invitrogen™
MA112059 |
50 Tests |
£414.00
50 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a monoclonal mixture of FITC-conjugated CD2 (clone T6.3, isotype IgG2a) and PE-conjugated CD20 (clone DFA-7, isotype IgG1).
CD2 is found on the majority of human T and NK cells. CD2 presents on 78+8% normal peripheral lymphocytes and 100% of thymocytes. This antibody detects CD2 antigen CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lamda light chains. It appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and malignant B cell proliferation. It is expressed in all B cell precursor leukemias.Specifications
| CD2/CD20 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Cocktail | |
| Liquid | |
| IgG1, IgG2a | |
| Human | |
| MS4A1 | |
| 2CD2 antigen; B1; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; CD2; CD2 molecule; CD20; CD20 antigen; CD20 receptor; CVID5; Erythrocyte receptor; LEU-16; leukocyte surface antigen Leu-16; LFA-2; LFA-3 receptor; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; MGC3969; MS4A1; MS4A2; Rosette receptor; S7; SRBC; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5 | |
| CD2, MS4A1 | |
| Primary | |
| Protein A/G |
| T6.3, DFA-7, DFA-7 | |
| Flow Cytometry | |
| FITC/PE | |
| Mouse | |
| RUO | |
| PBS with 0.2% carrier protein and 0.08% sodium azide | |
| P06729, P11836 | |
| 914, 931 | |
| T lymphocytes activated by mixed lymphocyte culture and the LB lymphoblastoid cell line. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title