missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (HuLy-m1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44908-0.1MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD2 Monoclonal specifically detects CD2 in Human, Feline samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Immunofluorescence.
Specifications
| CD2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| P06729 | |
| CD2 | |
| Human thymocytes | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| CD2 interacts through its amino-terminal domain with the extracellular domain of CD58 (also designated CD2 ligand) to mediate cell adhesion. CD2/CD58 binding can enhance antigen-specific T cell activation. CD2 is a transmembrane glycoprotein that is expressed on peripheral blood T lymphocytes, NK cells and thymocytes. CD58 is a heavily glycosylated protein with a broad tissue distribution in hematopoietic and other cells, including endothelium. Interaction between CD2 and its counter receptor LFA3 (CD58) on opposing cells optimizes immune system recognition, thereby facilitating communication between helper T lymphocytes and antigen-presenting cells, as well as between cytolytic effectors and target cells. | |
| Store at 4C. | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence | |
| HuLy-m1 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Mouse | |
| 50 kDa | |
| 0.1 mg | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 914 | |
| Human, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction