missing translation for 'onlineSavingsMsg'
Learn More

CD2 Antibody (3D1E3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£184.00 - £456.00

Specifications

Antigen CD2
Clone 3D1E3
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin, Microarray
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18031406
View Documents
Novus Biologicals
NBP2-61727
0.1 mL
£456.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18614947
View Documents
Novus Biologicals
NBP2-61727-0.025ml
0.025 mL
£184.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD2 Monoclonal specifically detects CD2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Microarray.
Specifications

Specifications

CD2
1 mg/mL
Monoclonal
Purified
Adaptive Immunity, Apoptosis, Immunology
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
CD2
IgG1
Protein G purified
3D1E3
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin, Microarray
Unconjugated
Mouse
Human, Mouse
914
Purified recombinant fragment of human CD2 (AA: 25-140) expressed in E. Coli.
Primary
39.4 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.