missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD1e Polyclonal specifically detects CD1e in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | CD1e |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | CD1e molecule, e polypeptide, FLJ17609, membrane-associated, thymocyte antigen CD1E |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human CD1e (NP_001036048.1). Peptide sequence GRLQLVCHVSGFYPKPVWVMWMRGEQEQRGTQRGDVLPNADETWYLRATL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?