missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Rat anti-Mouse, BUV563, Clone: 1B1, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 741287
This item is not returnable.
View return policy
Description
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD1d | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| Cd1d1; Cd1.1; Cd1a; Cd1d; Ly-38 | |
| Mouse Cd1.1 cDNA-transfected RMA-S mouse T lymphoma and L929 cells | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| 1B1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction