missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD1d Monoclonal Antibody (1B1), NovaFluor™ Red 685, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£151.00 - £294.00
Specifications
| Antigen | CD1d |
|---|---|
| Clone | 1B1 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089864
|
Invitrogen™
M049T02R02-A |
25 μg |
£151.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089617
|
Invitrogen™
M049T03R02-A |
100 μg |
£294.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow CytometrySpecifications
| CD1d | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AI747460; Antigen-presenting glycoprotein CD1d; antigen-presenting glycoprotein CD1d1; Cd1; CD1.1; CD1A; Cd1d; CD1D antigen; CD1D antigen, d polypeptide; CD1d molecule; CD1d.1; Cd1d1; CD1d1 antigen; CD1d1 molecule; differentiation antigen CD1-alpha-3; HMC class I antigen-like glycoprotein CD1D; Ly-38; R3; R3G1; T-cell surface glycoprotein CD1d; T-cell surface glycoprotein CD1d1; thymocyte antigen CD1D | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| 1B1 | |
| Flow Cytometry | |
| NovaFluor Red 685 | |
| Rat | |
| Mouse | |
| P11609 | |
| 12479 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Cd1d1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title