missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Antibody (1B1), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43461PE
This item is not returnable.
View return policy
Description
CD1d Monoclonal specifically detects CD1d in Mouse, Hamster samples. It is validated for Flow Cytometry.
Specifications
| CD1d | |
| Monoclonal | |
| PE | |
| antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| 1B1 | |
| Flow Cytometry | |
| CD1D | |
| The immunogen for this antibody was CD1d. | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Myeloid derived Suppressor Cell | |
| 912 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction