missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1c/BDCA-1 Antibody (OTI2F4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70345
This item is not returnable.
View return policy
Description
CD1c/BDCA-1 Monoclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CD1c/BDCA-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:200 - 1:1000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:150 | |
| BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
| Mouse | |
| 35.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI2F4 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| CD1C | |
| Full length human recombinant protein of human CD1C(NP_001756) produced in HEK293T cell. | |
| Affinity Purified | |
| Cancer, Immunology | |
| 911 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction