missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1b/c Antibody (B-B5) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18096JF525
This item is not returnable.
View return policy
Description
CD1b/c Monoclonal antibody specifically detects CD1b/c in Human samples. It is validated for Flow Cytometry
Specifications
| CD1b/c | |
| Monoclonal | |
| Janelia Fluor 525 | |
| BDCA1, CD1, CD1A, CD1B antigen, b polypeptide, CD1C antigen, c polypeptide, cortical thymocyte antigen CD1B, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R1, R7, T-cell surface glycoprotein CD1b, T-cell surface glycoprotein CD1c | |
| Thymus cells and Jurkat cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
| 0.1 mL | |
| CD Markers | |
| 910 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| B-B5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction