missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD1b Monoclonal antibody specifically detects CD1b in Human samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Antigen | CD1b |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 2E4 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_001755 |
| Gene Alias | CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b |
| Host Species | Mouse |
| Immunogen | CD1B (NP_001755.1, 19 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EHAFQGPTSFHVIQTSSFTNSTWAQTQGSGWLDDLQIHGWDSDSGTAIFLKPWSKGNFSDKEVAELEEIFRVYIFGFAREVQDFAGDFQMKY |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?