missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Rabbit anti-Human, Clone-C1A/1506R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54547-100UG
This item is not returnable.
View return policy
Description
CD1a Monoclonal specifically detects CD1a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.
Specifications
| CD1a | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Rabbit | |
| 49 kDa | |
| 100 μg | |
| Dendritic Cell Markers, Immunology | |
| 909 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| C1A/1506R | |
| Unconjugated | |
| PBS with No Preservative | |
| CD1A | |
| Recombinant human full-length CD1a protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| IgG κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur