missing translation for 'onlineSavingsMsg'
Learn More

CD1a Mouse anti-Human, FITC, Clone: HI149, BD

Mouse Monoclonal Antibody

£62.31 - £156.31

Specifications

Antigen CD1a
Clone HI149
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15890619
View Documents
BD Biosciences
560945
25 Tests
ONLINE EXCLUSIVE PRICE
£64.20
£62.31 / Each
Save £1.89
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15888448
View Documents
BD Biosciences
555806
100 Tests
ONLINE EXCLUSIVE PRICE
£163.00
£156.31 / Each
Save £6.69
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HI149 monoclonal antibody specifically binds to CD1a. CD1a is a type I transmembrane glycoprotein. The 49 kDa CD1a polypeptide is associated with β2-microglobulin. CD1a is expressed on cortical thymocytes, dendritic cells and Langerhans cells. CD1a has structural similarities to the MHC class I antigen, and plays a role in antigen presentation.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CD1a
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
HI149
Monoclonal
Mouse
Human
IgG1 κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.