missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Antibody (NA1/34-HLK), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63529
This item is not returnable.
View return policy
Description
CD1a Monoclonal specifically detects CD1a in Human, Canine samples. It is validated for Flow Cytometry.
Specifications
| CD1a | |
| Monoclonal | |
| 0.1 mg/mL | |
| Flow Cytometry 1:10-1:1000 | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| NB100-64852 recognizes the human CD1a cell surface glycoprotein, a 49kD molecule expressed in association with beta 2 microglobulin. CD1a is expressed strongly by cortical thymocytes, and also by Langerhans cells and interdigitating cells. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry | |
| NA1/34-HLK | |
| FITC | |
| P06126 | |
| CD1A | |
| Human thymocytes | |
| 0.1 mg | |
| Dendritic Cell Markers, Immunology | |
| 909 | |
| Human, Canine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction