missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD1a Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD1a Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-746-S

 View more versions of this product

Product Code. 12672237

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

At least five CD1 genes (CD1a, b, c, d, and e) have been identified. CD1 is expressed on cortical thymocytes, Langerhans cells, and dendritic cells. It is absent on mature peripheral blood T cells but intracytoplasmic expression is detected on activated T lymphocytes. CD1 proteins have been demonstrated to restrict T-cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation.

Host Species: Mouse

Clone: MTB1

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: External domain

Immunogen: Full external domain of CD1a

Molecular Weight: 49 kDa

Positive Control: Thymus

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD1a Ab-2
Monoclonal
Unconjugated
Full external domain of CD1a
1 mL
Hematopoietic
Human
IgG2a κ
Immunohistochemistry (Paraffin)
MTB1
Mouse
Thymus
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.