missing translation for 'onlineSavingsMsg'
Learn More

CD197 (CCR7) Rat anti-Human, PE, Clone: 3D12, BD

Rat Monoclonal Antibody

£65.89 - £259.26

Specifications

Antigen CD197
Clone 3D12
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15880679
View Documents
BD Biosciences
561008
25 Tests
ONLINE EXCLUSIVE PRICE
£68.50
£65.89 / Each
Save £2.61
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10686564
View Documents
BD Biosciences
552176
100 Tests
ONLINE EXCLUSIVE PRICE
£270.00
£259.26 / Each
Save £10.74
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The monoclonal antibody 3D12 reacts with the human CC chemokine receptor, CCR7. CCR7 (previously known as BLR-2, EBI-1 and CMKBR7), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CC chemokines, MIP-3β/Exodus 3/ELC/ CCL19 and 6Ckine/Exodus 2/SLC/TCA4/CCL21. It has been shown that CCR7 mRNA is expressed mainly in lymphoid tissues including spleen, lymph nodes and tonsil. CCR7 mRNA was also detected in peripheral T and B lymphocytes, in bone marrow and cord blood CD34-positive cells and mature dendritic cells. The human CCR7 gene, unlike other CC chemokine receptor genes, has been mapped to chromosome 17q12. The immunogen used to generate 3D12 hybridoma was the N-terminus as well as parts of the second extracellular loop of human CCR7 protein. The monoclonal antibody 3D12 recognizes an epitope mapping to the N-terminus of human CCR7.

Specifications

Specifications

CD197
Flow Cytometry
PE
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
CCR7, BLR-2, EBI-1, CMKBR7
3D12
Monoclonal
Rat
Human
Human CCR7 protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.