missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD197, CCR7 Rat anti-Human, BB515, Clone: 3D12, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 565869
This item is not returnable.
View return policy
Description
CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.
CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.
Specifications
| CD197, CCR7 | |
| Monoclonal | |
| Brilliant Blue 515 (BB515) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry | |
| 3D12 | |
| Vol. Per test: 5μL | |
| CCR7, BLR-2, EBI-1, CMKBR7 | |
| Human CCR7 protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction