missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD196 (CCR6), PE, clone: R6H1, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 12-1969-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This R6H1 monoclonal antibody reacts with CD196 (also known as CCR6), a seven transmembrane G protein-coupled receptor expressed on T, B, dendritic, natural killer, and Langerhans cells. This CC chemokine receptor uniquely binds MIP-3a/CCL20, a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CD196 is also involved in host defense and inflammation at epithelial sites. Furthermore, this receptor has been implicated in Th17 differentiation and CD4+FoxP3+ regulatory T cell development.Applications Reported: This R6H1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This R6H1 antibody has been pre-titrated and tested by flow cytometric analysis on normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
CD196 (CCR6) | |
Monoclonal | |
5μl/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P51684 | |
BN-1, CKR-L3, CKR6, CKRL3, CMKBR6, DCR2, DRY-6, GPR-CY4, GPR29, GPRCY4, STRL22 | |
Murine | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
R6H1 | |
PE | |
CCR6 | |
Conjugated | |
CCR6 | |
IgG1 | |
100 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
1235 |
For Research Use Only.