Learn More
CD196 (CCR6) Mouse anti-Human, PE, Clone: 11A9, BD
Mouse Monoclonal Antibody
£102.63 - £290.95
Specifications
| Antigen | CD196 |
|---|---|
| Clone | 11A9 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15870689
|
BD Biosciences
561019 |
25 μg |
£107.00 £102.63 / Each Save £4.37 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15879738
|
BD Biosciences
559562 |
0.2 mg |
£303.00 £290.95 / Each Save £12.05 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 11A9 monoclonal antibody specifically binds to CCR6. CCR6 is a seven-transmembrane, G-protein-coupled, glycoprotein receptor that is a member of the beta chemokine receptor family. CCR6 was clustered as CD196 in the 7th HLDA Workshop. The human CCR6 gene has been mapped to chromosome 6q27. CCR6 is a receptor for the CC chemokine CCL20/MIP-3alpha/LARC/Exodus and also binds with lower affinity to and mediates responses to beta-defensin2/hBD-2. CCR6 is predominantly expressed by B lymphocytes, certain subsets of effector and memory T cells and by immature dendritic cells but not by monocytes, NK cells, or granulocytyes. Skin-homing CLA (Cutaneous Lymphocyte Antigen)-positive memory T cells, Th1 cells, regulatory T cells and IL-17A-producing Th17 cells predominantly express high levels of CCR6. CCR6 mediates the trafficking of T, B, and dendritic cells to epithelial sites near the skin and mucosal surfaces during inflammatory and immunological responses. An N-terminal peptide of human CCR6 was used as an immunogen to generate the 11A9 hybridoma. The 11A9 antibody does not cross-react with human CCR1, CCR2, CCR3, CCR4, CCR5, CCR7, CCR8, CCR9, CXCR1, CXCR2, CXCR3, CXCR4 and CXCR5 receptors. This antibody is NOT a neutralizing antibody.
Specifications
| CD196 | |
| 0.2mg/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human CD196/CCR6 Peptide | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 11A9 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| BN-1; C-C chemokine receptor type 6; C-C CKR-6; CC-CKR-6; CCR-6; CD196 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.