missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD193 Rat anti-Mouse, BUV737, Clone: 83103, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 741699
This item is not returnable.
View return policy
Description
CD193 Monoclonal antibody specifically detects CD193 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD193 | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| CCR3; CCR-3; chemokine (C-C) receptor 3; CC-CKR3; C-C CKR-3; CKR3; CC-CKR-3 | |
| Mouse CCR3 transfected Cells Transfected Cell Line | |
| RUO | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 83103 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering