missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, Biotin, Clone: 1D3, enQuireBio™
Rat Monoclonal Antibody
£62.45 - £219.00
Specifications
Antigen | CD19 |
---|---|
Clone | 1D3 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961758
|
EnquireBio
QAB28-B-25ug |
25 μg |
£62.45
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951758
|
EnquireBio
QAB28-B-100ug |
100 μg |
£155.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15971758
|
EnquireBio
QAB28-B-500ug |
500 μg |
£219.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD19 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
12478 | |
IgG2a κ | |
Protein A/G Purified |
1D3 | |
Flow Cytometry | |
Biotin | |
RUO | |
NP_033974 | |
Cd19 | |
Primary | |
Store at 2-4°C. |