missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse, anti-Human, Cardinal Red, Clone: HIB19, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CR-65110-100TESTS
This item is not returnable.
View return policy
Description
The CD19 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD19. This antibody recognizes human antigen. The CD19 antibody has been validated for the following applications: FC analysis. This antibody is cardinal red™.
Specifications
| CD19 | |
| HIB19 | |
| Cardinal Red | |
| Mouse | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 5 μL/Test | |
| B lymphocyte antigen CD19, B4, CD19, CD19 molecule, Differentiation antigen CD19, T cell surface antigen Leu 12 | |
| 100 Tests | |
| 930 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction