missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse, anti-Human, Atlantic Blue, Clone: HIB19, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech AB-65110-100TESTS
This item is not returnable.
View return policy
Description
The CD19 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD19. This antibody recognizes human antigen. The CD19 antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.
Specifications
| CD19 | |
| HIB19 | |
| Atlantic Blue | |
| Mouse | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 5 μL/Test | |
| B lymphocyte antigen CD19, B4, CD19, CD19 molecule, Differentiation antigen CD19, T cell surface antigen Leu 12 | |
| 100 Tests | |
| 930 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion