missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse anti-Human, APC-R700, Clone: HIB19, BD Horizon™
Mouse Monoclonal Antibody
£190.92 - £342.74
Specifications
| Antigen | CD19 |
|---|---|
| Clone | HIB19 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-R700 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15883999
|
BD Biosciences
564978 |
25 Tests |
£200.00 £190.92 / Each Save £9.08 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15873999
|
BD Biosciences
564977 |
100 Tests |
£357.00 £342.74 / Each Save £14.26 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometry.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometry.Specifications
| CD19 | |
| Flow Cytometry | |
| APC-R700 | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed. |
| HIB19 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B4; B-lymphocyte antigen CD19; Leu-12 | |
| Primary | |
| Vol. Per test: 5μL |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title