Learn More
Invitrogen™ CD19 Monoclonal Antibody (eBio1D3 (1D3)), FITC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£75.25 - £390.00
Specifications
| Antigen | CD19 |
|---|---|
| Clone | eBio1D3 (1D3) |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15288039
|
Invitrogen™
11-0193-81 |
50 μg |
£75.25
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15219185
![]() |
Invitrogen™
11-0193-82 |
100 μg |
£138.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15298039
|
Invitrogen™
11-0193-85 |
500 μg |
£234.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15208049
|
Invitrogen™
11-0193-86 |
1 mg |
£390.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. Applications Reported: This eBio1D3 (1D3) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio1D3 (1D3) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lambda light chains. CD19 appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and malignant B cell proliferation, and is expressed in all B cell precursor leukemias. Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. CD19 is a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. Besides being a signal-amplifying coreceptor for the B cell receptor (BCR), CD19 can also signal independently of BCR co-ligation and is a central regulatory component upon which multiple signaling pathways converge. Mutation of the CD19 gene results in hypogammaglobulinemia, whereas CD19 overexpression causes B cell hyperactivity.Specifications
| CD19 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
| Cd19 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Cd19 |
| eBio1D3 (1D3) | |
| Flow Cytometry | |
| FITC | |
| Rat | |
| Mouse | |
| P25918 | |
| 12478 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.