Learn More
CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£84.75 - £151.00
Specifications
Antigen | CD19 |
---|---|
Clone | eBio1D3 (1D3) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15391690
|
Affymetrix eBioscience
48-0193-80 |
25 μg |
£84.75
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15301700
|
Affymetrix eBioscience
48-0193-82 |
100 μg |
£151.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. Applications Reported: This eBio1D3 (1D3) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio1D3 (1D3) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD19 Monoclonal antibody specifically detects CD19 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD19 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Cd19 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd19 |
eBio1D3 (1D3) | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
P25918 | |
12478 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.