missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (FMC63) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-52716-0.2MG
This item is not returnable.
View return policy
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| CD19 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| CD19 | |
| Human prolymphocytic leukaemia cell line JVM3. | |
| 0.2 mg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 930 | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, CyTOF | |
| FMC63 | |
| Flow Cytometry 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Binds human CD19. | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction