missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD19 Monoclonal antibody specifically detects CD19 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, In vitro Assay, CyTOF
Specifications
Specifications
| Antigen | CD19 |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, In vitro Assay, CyTOF |
| Classification | Monoclonal |
| Clone | CB19 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 |
| Host Species | Mouse |
| Immunogen | Intact normal human B cell cells were used as immunogen to generate the CB19 clone. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?