missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (CB19) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80634
This item is not returnable.
View return policy
Description
CD19 Monoclonal specifically detects CD19 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, In vitro assay, CyTOF-ready.
Specifications
| CD19 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot reported in scientific literature (PMID 17327405), Flow Cytometry : 0.2 ug/10^6 cells, Immunocytochemistry/Immunofluorescence : 1:25-1:100, In vitro assay reported in scientific literature (PMID 9576485), CyTOF-ready | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse reactivity reported in scientific literature (PMID: 27532872). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, In vitro Assay, CyTOF | |
| CB19 | |
| Unconjugated | |
| PBS | |
| CD19 | |
| Intact normal human B cell cells were used as immunogen to generate the CB19 clone. | |
| 0.1 mg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 930 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction