missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
Specifications
| Antigen | CD19 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | CAT-13.1E10 |
| Conjugate | DyLight 755 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 |
| Host Species | Mouse |
| Immunogen | This antibody was raised by immunising mice against CD19. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?