Learn More
CD18 (LFA-1 beta) Monoclonal Antibody (M18/2), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 12-0181-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The M18/2 monoclonal antibody reacts with mouse CD18, the 90-95 kDa integrin beta 2 glycoprotein. CD18 is the common beta subunit that non-covalently associates with CD11a (alpha L), CD11b (alpha M) and CD11c (alpha X) to form LFA-1, Mac-1 and gp150/95. Heterodimers of CD18 and the a subunits are expressed broadly by all leukocytes with NK and T cells showing higher density of surface expression. Through the interactions of LFA-1, Mac-1 and gp150/95 with their counter-receptors, CD18 is involved in cellular adhesion and the immune response. Applications Reported: The M18/2 antibody has been reported for use in flow cytometric analysis. Applications Tested: The M18/2 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.
CD18 integrin beta 2 subunit is a 90 kDa type I transmembrane protein expressed on all leucocytes. CD18 can combine with integrin molecules CD11a-c to form heterodimers at the cell surface, and these heterodimers are known to participate in the process of cell adhesion as well as cell-surface mediated signaling. CD18 forms heterodimers with four types of CD11 molecule to constitute leukocyte (beta2) integrins: alphaLbeta2 (CD11a/CD18, LFA-1), alphaMbeta2 (CD11b/CD18, Mac-1, CR3), alphaXbeta2 (CD11c/CD18) and alphaDbeta2 (CD11d/CD18). In most cases, the response mediated by the integrin is a composite of the functions of its individual subunits, and these integrins are essential for proper leukocyte migration, mediating intercellular contacts. Absence of CD18 leads to leukocyte adhesion deficiency-1, severe reduction of CD18 expression leads to the development of a psoriasiform skin disease. CD18 is also a target of Mannheimia (Pasteurella) haemolytica leukotoxin and is sufficient to mediate leukotoxin-mediated cytolysis. Defects in the CD18 gene are the cause of leukocyte adhesion deficiency type I (LAD1). Two transcript variants encoding the CD18 protein have been identified.Specifications
CD18 (LFA-1 beta) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P11835 | |
Itgb2 | |
Affinity chromatography | |
RUO | |
16414 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
M18/2 | |
PE | |
Itgb2 | |
Cd18, LAD, LCAMB, Lfa1, MF17, Integrin beta-2 subunit, ITGB2, cluster designation 18 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.