Learn More
Invitrogen™ CD179b Recombinant Rabbit Monoclonal Antibody (8H16L35)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702901
Description
This antibody is predicted to react with Monkey, Cat, Pig, Rat. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
The preB cell receptor, CD179b, is found on the surface of proB and preB cells, where it is involved in transduction of signals for cellular proliferation and differentiation from the proB cell to the preB cell stage, allelic exclusion at the Ig heavy chain gene locus, and promotion of Ig light chain gene rearrangements. The preB cell receptor is composed of a membrane-bound Ig mu heavy chain in association with a heterodimeric surrogate light chain. This gene encodes one of the surrogate light chain subunits and is a member of the immunoglobulin gene superfamily. Mutations in this gene can result in B cell deficiency and agammaglobulinemia, an autosomal recessive disease in which few or no gamma globulins or antibodies are made. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| CD179b | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P15814 | |
| IGLL1 | |
| Peptides corresponding to human CD179b [1) aa93-aa112 2) aa152-aa167]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 8H16L35 | |
| Unconjugated | |
| IGLL1 | |
| 14.1; AGM2; BB139905; CD179 antigen-like family member B; CD179b; CD179b antigen; ig lambda-5; IGL1; IGL5; Igl-5; IGLJ IGLL; IGLJ14.1; Igll; Igll1; IGO; IGVPB; immunoglobulin lambda chain 5; immunoglobulin lambda like polypeptide 1; immunoglobulin lambda-like polypeptide 1; immunoglobulin lambda-like polypeptide 5; Immunoglobulin omega polypeptide; immunoglobulin omega polypeptide chain; immunoglobulin-related 14.1 protein; immunoglobulin-related protein 14.1; lambda 5; lambda5; Pre-B lymphocyte-specific protein-2; VPREB2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3543 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.