Learn More
CD178 / Fas ligand Mouse, APC, Clone: NOK-1, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio QAB85-APC-100Tests
 View more versions of this product
Product Code. 15922568
  • £254.10 / 100 tests
Specifications
CD178 / Fas ligand
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
FASLG
Protein A/G Purified
RUO
356
IgG1 κ
Flow Cytometry
NOK-1
APC
NP_001289675
Mouse
100 Tests
Primary
Store at 2-4°C.