missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD177 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16310785
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16310785 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16310785 Supplier Invitrogen™ Supplier No. PA583827

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: CQFGTVQHVW KVSDLPRQWT PKNTSCDSGL GCQDTLMLIE SGPQVSLVLS KGCTEAKDQE PRVTEHRMGP GLSLISYTFV CRQEDFCNNL VNSLP Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 56%.

CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD177
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CD177
Gene Accession No. Q8N6Q3
Gene Alias 1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181
Gene Symbols CD177
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CD177. Recombinant protein control fragment (Product #RP-106130).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.