missing translation for 'onlineSavingsMsg'
Learn More

CD173 Mouse anti-Human, Clone: A51-B/A6, Novus Biologicals™

Product Code. 18342075 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342075 20 μg 20µg
18330994 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18342075 Supplier Novus Biologicals Supplier No. NBP31393120UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD173
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A51-B/A6
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry 1-2 ug/ml, Immunofluorescence 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P16442
Gene Alias H2, Type 2 H blood group antigen
Host Species Mouse
Immunogen Human breast cancer MCF-7 cells
Purification Method PEG purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.