missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD173 Antibody (BRIC231), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62120CL1
This item is not returnable.
View return policy
Description
CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Flow Cytometry,Haemagglutination,Immunofluorescence
Specifications
| CD173 | |
| Monoclonal | |
| CoraFluor 1 | |
| H2, Type 2 H blood group antigen | |
| Human erythroleukemic cell line (HEL) established from a 30 year old patient with relapsed erythroleukemia following treatment for Hodgkin lymphoma | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Hemagglutination Assay, Immunofluorescence | |
| BRIC231 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction