missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD173 Antibody (A51-B/A6) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14046JF525
This item is not returnable.
View return policy
Description
CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CD173 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| H2, Type 2 H blood group antigen | |
| Human breast cancer MCF-7 cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgA |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| A51-B/A6 | |
| 50mM Sodium Borate | |
| Mouse | |
| PEG purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction