missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD173 Antibody (A51-B/A6), DyLight 405, Novus Biologicals™
Description
CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CD173 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | A51-B/A6 |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | H2, Type 2 H blood group antigen |
| Host Species | Mouse |
| Immunogen | Human breast cancer MCF-7 cells |
| Purification Method | PEG purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?