missing translation for 'onlineSavingsMsg'
Learn More

CD173 Antibody (A46-B/B10) - Azide and BSA Free, Novus Biologicals™

Product Code. 18372283 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372283 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372283 Supplier Novus Biologicals Supplier No. NBP314040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Hemagglutination, Binding Inhibition
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD173
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Hemagglutination Assay, Inhibition Assays
Classification Monoclonal
Clone A46-B/B10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 2 to 4μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Hemagglutination, Binding Inhibition
Formulation 10mM PBS without Preservative
Gene Alias H2, Type 2 H blood group antigen
Host Species Mouse
Immunogen Human breast cancer MCF-7 cells
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.