Learn More
CD171 Monoclonal Antibody (eBio5G3 (5G3)), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 17-1719-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
The monoclonal antibody eBio5G3 recognizes CD171 also known as neural cell adhesion molecule L1. CD171 is a member of the Ig superfamily containing 6 extracellular Ig domains and five fibronectin type III-like repeats. CD171 has been shown to function as a cell adhesion molecule mediating homotypic and heterotypic cell-cell interactions in neuronal myelination, neurite outgrowth and regeneration. Expression of CD171 has been found on monocytes and mature monocytic-derived and follicular DCs, a minor subset of lymphocytes in addition to that found on neuronal tissue and some tumor cells lines. Expression of CD171 on tumors is thought to contribute to tumor progression. Epitope of eBio5G3 is in amino-terminal Ig-like domain. This eBio5G3 (5G3) antibody has been pre-titrated and tested by flow cytometric analysis of the tumor cell line Panc-1. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 105 to 108 cells/test. Filtration: 0.2 μm post-manufacturing filtered.
CD171 Monoclonal antibody specifically detects CD171 in Human samples. It is validated for Flow Cytometry.Specifications
CD171 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% Sodium Azide; pH 7.2 | |
P32004 | |
L1CAM | |
Affinity chromatography | |
RUO | |
3897 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBio5G3 (5G3) | |
APC | |
L1CAM | |
antigen identified by monoclonal antibody R1; CAML1; CD 171; CD171; CD171 molecule; HSAS; HSAS1; Hyd; L1; L1 cell adhesion molecule; L1CAM; MASA; MIC5; N-CAM L1; NCAML1; N-CAML1; N-CAM-L1; NCAM-L1; Nerve-growth factor-inducible large external glycoprotein; neural cell adhesion molecule L1; NILE; S10; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM; SPG1 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
For Research Use Only.