missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD170 Mouse anti-Human, BV650, Clone: 1A5/CD170 (also known as 1A5), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 749695
This item is not returnable.
View return policy
Description
CD170 Monoclonal antibody specifically detects CD170 in Human samples. It is validated for Flow Cytometry
Specifications
| CD170 | |
| Monoclonal | |
| Brilliant Violet 650 | |
| SIGLEC-5; SIGLEC5; CD33 antigen-like 2; CD33L2; obesity-binding protein 2; OBBP2 | |
| Human Siglec-5 Recombinant Protein | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 1A5/CD170 (also known as 1A5) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV650 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction